Assay Details
| Gene Symbol: Dclre1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018831.4 | NP_061301.3 | 6 - 7 | 3128 | 67 | |
| GenBank mRNA | AF241240.1 | - | 5 - 6 | 2593 | 67 | |
| AK029794.1 | - | 5 - 6 | 2797 | 67 | ||
| AK220214.1 | - | 5 - 6 | 2350 | 67 | ||
| BC072667.1 | - | 4 - 5 | 3726 | 30629417 | 67 | |
| BC076617.1 | - | 6 - 7 | 3128 | 67 | ||
More Information
Set Membership: |
|

Dclre1a
Genomic Map