| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00017176.mm.1 |
NM_001177985.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017176.mm.1 |
NM_001177985.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017176.mm.1 |
NM_001177986.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017176.mm.1 |
NM_001177986.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017176.mm.1 |
NM_001177987.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017176.mm.1 |
NM_001177987.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017176.mm.1 |
NM_001177988.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017176.mm.1 |
NM_001177988.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017176.mm.1 |
NM_019831.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017176.mm.1 |
NM_019831.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017176.mm.1 |
TC0X00002721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00017190.mm.1 |
NM_001177985.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017190.mm.1 |
NM_001177985.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017190.mm.1 |
NM_001177986.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017190.mm.1 |
NM_001177986.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017190.mm.1 |
TC0X00002721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00017185.mm.1 |
NM_001177985.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017185.mm.1 |
NM_001177985.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017185.mm.1 |
NM_001177986.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017185.mm.1 |
NM_001177986.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017185.mm.1 |
TC0X00002721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00017197.mm.1 |
NM_001177987.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017197.mm.1 |
NM_001177987.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00017197.mm.1 |
TC0X00002721.mm.1 |
MTA TC |