Assay Details
| Gene Symbol: Noc3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021315.2 | NP_067290.2 | 7 - 8 | 993 | 80 | |
| GenBank mRNA | AB077991.1 | - | 7 - 8 | 956 | 80 | |
| AK134051.1 | - | 7 - 8 | 951 | 80 | ||
| AK168944.1 | - | 7 - 8 | 994 | 80 | ||
| BC031132.1 | - | 7 - 8 | 956 | 4954951 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Noc3l
Genomic Map