Assay Details
| Gene Symbol: Rqcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021383.5 | NP_067358.1 | 810 | 131 | ||
| GenBank mRNA | AF221849.1 | - | 4 - 5 | 432 | 131 | |
| AK005025.2 | - | 4 - 5 | 812 | 131 | ||
| BC050898.1 | - | 4 - 5 | 611 | 6401907 | 131 | |
| BC051948.1 | - | 649 | 5708192 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rqcd1
Genomic Map