Assay Details
| Gene Symbol: Sdhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023374.3 | NP_075863.2 | 1 - 2 | 207 | 143 | |
| GenBank mRNA | AK003052.1 | - | 1 - 2 | 101 | 143 | |
| AK003533.1 | - | 1 - 2 | 91 | 143 | ||
| AK009660.1 | - | 1 - 2 | 101 | 143 | ||
| AK153045.1 | - | 1 - 2 | 84 | 143 | ||
| AK169252.1 | - | 1 - 2 | 98 | 143 | ||
| BC013509.1 | - | 1 - 2 | 102 | 4225025 | 143 | |
| BC018580.1 | - | 1 - 2 | 66 | 143 | ||
| BC051934.1 | - | 1 - 2 | 207 | 5688898 | 143 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sdhb
Genomic Map