Assay Details
| Gene Symbol: Sdhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023374.3 | NP_075863.2 | 3 - 4 | 426 | 114 | |
| GenBank mRNA | AK003052.1 | - | 3 - 4 | 320 | 114 | |
| AK003533.1 | - | 3 - 4 | 310 | 114 | ||
| AK009660.1 | - | 3 - 4 | 320 | 114 | ||
| AK153045.1 | - | 3 - 4 | 303 | 114 | ||
| AK169252.1 | - | 3 - 4 | 317 | 114 | ||
| AK169839.1 | - | 3 - 4 | 319 | 114 | ||
| AK196072.1 | - | 1 - 2 | 83 | 114 | ||
| BC013509.1 | - | 3 - 4 | 321 | 4225025 | 114 | |
| BC018580.1 | - | 3 - 4 | 285 | 114 | ||
| BC051934.1 | - | 3 - 4 | 426 | 5688898 | 114 | |
| DQ403000.1 | - | 2 - 3 | 217 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sdhb
Genomic Map