Assay Details
| Gene Symbol: Smg8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024262.1 | NP_077224.1 | 3 - 4 | 2802 | 104 | |
| GenBank mRNA | AK004714.1 | - | 3 - 4 | 2795 | 104 | |
| AK031106.1 | - | 3 - 4 | 2776 | 104 | ||
| AK035089.1 | - | 3 - 4 | 1769 | 104 | ||
| BC020005.1 | - | 3 - 4 | 2802 | 3981459 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27992496 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27992492 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27992491 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27992495 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27992494 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27992493 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P238902 | BC002247.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P238902 | BC020005.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P238902 | AK031106.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P238902 | AK035089.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P238902 | NM_024262.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P238902 | BC002247.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P238902 | BC020005.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P238902 | AK031106.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P238902 | AK035089.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P238902 | NM_024262.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 312437 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527251 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322179 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 393301 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 585558 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906155 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 10680 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116717 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P238902 | BC002247.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P238902 | BC020005.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P238902 | AK031106.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P238902 | AK035089.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P238902 | NM_024262.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P238902 | BC002247.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P238902 | BC020005.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P238902 | AK031106.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P238902 | AK035089.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P238902 | NM_024262.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27992500 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116717 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07430258 | - | - |
| Whole Mouse Genome G4122A | A_52_P238902 | BC002247.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P238902 | BC020005.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P238902 | AK031106.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P238902 | AK035089.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P238902 | NM_024262.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P238902 | BC002247.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P238902 | BC020005.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P238902 | AK031106.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P238902 | AK035089.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P238902 | NM_024262.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118018 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116717 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09691308 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08993974 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09691321 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27992494 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27992495 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27992491 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27992492 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27992493 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27992496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27992414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09024146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09273594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09681734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09513678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09691333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09691324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09538345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09721122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27992447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09128277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09734853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09513677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09587461 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10389604 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389603 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389602 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389605 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427737_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10389601 | - | - |
| Mouse Genome 430 2.0 Array | 1427737_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1427737_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74133_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74133_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100031851.mm.1 | NM_024262.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031851.mm.1 | NM_024262.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031851.mm.1 | TC1100003469.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427737_a_at | - | - |
| Mouse Expression Set 430 | 1427737_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427737_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427737_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427737_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427737_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 746054 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6590450 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6590450 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006590450 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2696126 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2696126 | - | - |
| mouse-6 v1.1 (Illumina) | scl39767.4.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6590450 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39767.4.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6590450 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2696126 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3031706 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3105117 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3105117 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3031706 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2696126 | - | - |

Smg8
Genomic Map