Assay Details
| Gene Symbol: Mrpl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025570.2 | NP_079846.1 | 1 - 2 | 125 | 72 | |
| GenBank mRNA | AB049645.1 | - | 1 - 2 | 98 | 72 | |
| AK004053.1 | - | 1 - 2 | 110 | 72 | ||
| AK011341.1 | - | 1 - 2 | 126 | 72 | ||
| AK015199.1 | - | 109 | 72 | |||
| AK168665.1 | - | 1 - 2 | 99 | 72 | ||
| BC039985.1 | - | 1 - 2 | 95 | 1244957 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440683 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440681 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440682 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440680 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440684 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P413698 | AK004053.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P413698 | AK011341.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P413698 | BC039985.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P413698 | NM_025570.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P413698 | AK004053.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P413698 | AK011341.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P413698 | BC039985.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P413698 | NM_025570.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 483718 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 69924 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894471 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 183797 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 92639 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105956 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P413698 | AK004053.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P413698 | AK011341.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P413698 | BC039985.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P413698 | NM_025570.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P413698 | AK004053.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P413698 | AK011341.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P413698 | BC039985.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P413698 | NM_025570.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P413698 | AK004053.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P413698 | AK011341.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P413698 | BC039985.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P413698 | NM_025570.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P413698 | AK004053.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P413698 | AK011341.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P413698 | BC039985.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P413698 | NM_025570.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440694 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440702 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440709 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23440680 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105956 | - | - |
| Whole Mouse Genome G4122A | A_51_P413698 | AK004053.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P413698 | AK011341.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P413698 | BC039985.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P413698 | NM_025570.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P413698 | AK004053.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P413698 | AK011341.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P413698 | BC039985.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P413698 | NM_025570.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128800 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105956 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03228411 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03587792 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03587791 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03228416 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03228410 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03228417 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440684 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440681 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440680 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440682 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23440683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04095664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03228416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03587791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04043535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04027356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03587793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03224159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03626817 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03224157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03608840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03334481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03228417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03334477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03334476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440674 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04095673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03950010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03228410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03512210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23440697 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10511154 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511150 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511151 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511153 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511152 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416093_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10511149 | - | - |
| Mouse Genome 430 2.0 Array | 1416093_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1416093_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66448_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66448_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400017422.mm.1 | NM_025570.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017422.mm.1 | NM_025570.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017422.mm.1 | TC0400002091.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400017415.mm.1 | NM_025570.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017415.mm.1 | NM_025570.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017415.mm.1 | TC0400002091.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400017420.mm.1 | NM_025570.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017420.mm.1 | NM_025570.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017420.mm.1 | TC0400002091.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416093_PM_a_at | - | - |
| Mouse Expression Set 430 | 1416093_a_at | - | - |
| Murine Genome U74v2 Set | 94875_at | - | - |
| Murine Genome U74v2 Set | 94875_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416093_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416093_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416093_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416093_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 472226 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2640494 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002640494 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2665316 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2665316 | - | - |
| mouse-6 v1.1 (Illumina) | scl066448.4_32-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2640494 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl066448.4_32-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2640494 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2665316 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2665316 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066448.4_32-S | - | - |

Mrpl20
Genomic Map