Assay Details
| Gene Symbol: Shisa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025858.3 | NP_080134.1 | 2 - 3 | 353 | 81 | |
| GenBank mRNA | AF520699.1 | - | 2 - 3 | 335 | 81 | |
| AK009225.1 | - | 2 - 3 | 293 | 81 | ||
| AK033545.1 | - | 2 - 3 | 313 | 81 | ||
| AK045368.1 | - | 2 - 3 | 283 | 81 | ||
| AK150460.1 | - | 2 - 3 | 318 | 81 | ||
| AK151632.1 | - | 2 - 3 | 318 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Shisa5
Genomic Map