Assay Details
| Gene Symbol: Mrps18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025878.1 | NP_080154.1 | 5 - 6 | 424 | 65 | |
| GenBank mRNA | AB049954.1 | - | 5 - 6 | 448 | 65 | |
| AK002554.1 | - | 2 - 3 | 169 | 65 | ||
| AK010250.1 | - | 5 - 6 | 431 | 65 | ||
| AK076033.1 | - | 2 - 3 | 169 | 65 | ||
| AK076044.1 | - | 2 - 3 | 169 | 65 | ||
| AK088540.1 | - | 5 - 6 | 420 | 65 | ||
| AK166597.1 | - | 5 - 6 | 445 | 65 | ||
| AK167067.1 | - | 5 - 6 | 425 | 65 | ||
| BC021752.1 | - | 5 - 6 | 424 | 5136720 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrps18b
Genomic Map