Assay Details
| Gene Symbol: Gatad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026033.2 | NP_080309.2 | 4 - 5 | 928 | 80 | |
| GenBank mRNA | AB047921.1 | - | 4 - 5 | 909 | 80 | |
| AK078416.1 | - | 4 - 5 | 873 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23446280 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23446279 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23446283 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23446277 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23446281 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23446282 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P119247 | AB047921.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P119247 | AK012921.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P119247 | AK078416.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P119247 | NM_026033.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985559 | BC019449.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 717929 | AK138923.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 717929 | D90205.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 717929 | NM_008370.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 395030 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 126854 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 359290 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 800074 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344581 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 54031 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134586 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985559 | BC019449.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P119247 | AB047921.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P119247 | AK012921.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P119247 | AK078416.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P119247 | NM_026033.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446306 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446314 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446307 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134586 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05466489 | - | - |
| Whole Mouse Genome G4122A | A_52_P429320 | AK213660.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P429320 | AB047921.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P429320 | AK012921.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P429320 | AK078416.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P429320 | NM_026033.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137375 | BE947732.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137375 | AK081359.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137375 | AB047921.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137375 | NM_026033.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P455982 | BC019449.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P455982 | AB047921.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P455982 | AK012921.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P455982 | AK078416.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P455982 | NM_026033.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101954 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119247 | AB047921.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119247 | AK012921.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119247 | AK078416.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119247 | NM_026033.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134586 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04668152 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04668149 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04396606 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04396605 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04105067 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04668147 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04668154 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23446282 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23446279 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23446281 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23446280 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23446283 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23446277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04900384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446259 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04325455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446262 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446273 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04215340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04526672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446268 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04232248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04929800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446241 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04900387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04499787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04396606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446256 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04668150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04499786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446274 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10527891 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10527892 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10527894 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10527887 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10527893 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10527889 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10527895 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425599_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452085_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10527888 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10527886 | - | - |
| Mouse Genome 430 2.0 Array | 1452085_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1425599_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1425599_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1452085_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67210_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67210_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500017360.mm.1 | NM_026033.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017360.mm.1 | NM_026033.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017360.mm.1 | TC0500001887.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500017358.mm.1 | NM_026033.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017358.mm.1 | NM_026033.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017358.mm.1 | TC0500001887.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500017359.mm.1 | NM_026033.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017359.mm.1 | NM_026033.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017359.mm.1 | TC0500001887.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500017362.mm.1 | NM_026033.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017362.mm.1 | NM_026033.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017362.mm.1 | TC0500001887.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500017361.mm.1 | NM_026033.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017361.mm.1 | NM_026033.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017361.mm.1 | TC0500001887.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500017357.mm.1 | NM_026033.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017357.mm.1 | NM_026033.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017357.mm.1 | TC0500001887.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425599_a_at | - | - |
| Mouse Expression Set 430 | 1452085_PM_at | - | - |
| Mouse Expression Set 430 | 1425599_PM_a_at | - | - |
| Mouse Expression Set 430 | 1452085_at | - | - |
| Murine Genome U74v2 Set | 165455_at | - | - |
| Murine Genome U74v2 Set | 165455_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452085_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452085_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425599_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425599_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452085_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425599_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425599_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452085_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 602994 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 940170 | - | - |
| MouseWG-6 v2 Expression BeadChip | 2630176 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2630176 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 940170 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002630176 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000940170 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1259903 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2594205 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2594205 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1259903 | - | - |
| mouse-6 v1.1 (Illumina) | scl067210.1_15-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0067210.1_10-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2630176 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 940170 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067210.1_15-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0067210.1_10-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 940170 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2630176 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1259903 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2982097 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2594205 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2982097 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2594205 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1259903 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067210.1_15-S | - | - |

Gatad1
Genomic Map