Assay Details
| Gene Symbol: Poldip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026389.3 | NP_080665.1 | 2 - 3 | 261 | 76 | |
| GenBank mRNA | AK196491.1 | - | 2 - 3 | 124 | 76 | |
| BC006833.1 | - | 2 - 3 | 261 | 3597851 | 76 | |
| BC012879.1 | - | 2 - 3 | 273 | 4166791 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27943792 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P163928 | BC006833.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P163928 | BC012879.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P163928 | AK165703.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P163928 | NM_026389.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P163928 | AK003702.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P163928 | BC006833.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P163928 | BC012879.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P163928 | AK165703.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P163928 | NM_026389.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P163928 | AK003702.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 694887 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 616975 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 99211 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 891475 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 746322 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 345677 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 128207 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158393 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170653 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P163928 | BC006833.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P163928 | BC012879.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P163928 | AK165703.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P163928 | NM_026389.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P163928 | AK003702.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P163928 | BC006833.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P163928 | BC012879.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P163928 | AK165703.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P163928 | NM_026389.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P163928 | AK003702.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943801 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943792 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943808 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943822 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07408532 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170653 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158393 | - | - |
| Whole Mouse Genome G4122A | A_52_P163928 | BC006833.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P163928 | BC012879.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P163928 | AK165703.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P163928 | NM_026389.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P163928 | AK003702.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P163928 | BC006833.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P163928 | BC012879.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P163928 | AK165703.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P163928 | NM_026389.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P163928 | AK003702.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208637 | BC006833.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208637 | BC012879.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140834 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158393 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170653 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09624199 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09624200 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09624201 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27943792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09088789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09396541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09088800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943826 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943824 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09624199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09088801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09278475 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10379210 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379208 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379206 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379213 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379205 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379212 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379209 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379211 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379207 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379214 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450727_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422732_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10379204 | - | - |
| Mouse Genome 430 2.0 Array | 1450727_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1422732_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1450727_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1422732_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67811_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67811_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100010858.mm.1 | NM_026389.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010858.mm.1 | NM_026389.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010858.mm.1 | TC1100001164.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422732_at | - | - |
| Mouse Expression Set 430 | 1450727_a_at | - | - |
| Mouse Expression Set 430 | 1422732_PM_at | - | - |
| Mouse Expression Set 430 | 1450727_PM_a_at | - | - |
| Murine Genome U74v2 Set | 99636_at | - | - |
| Murine Genome U74v2 Set | 99636_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422732_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450727_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422732_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450727_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450727_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450727_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422732_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422732_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 566105 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2650139 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100450059 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002650139 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2760263 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2760263 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1242518 | - | - |
| mouse-6 v1.1 (Illumina) | scl41198.10_516-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100450059 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2650139 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl41198.10_516-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100450059 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2650139 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2760263 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2760263 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2543314 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl41198.10_516-S | - | - |

Poldip2
Genomic Map