| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900027808.mm.1 |
NM_026917.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027808.mm.1 |
NM_026917.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027808.mm.1 |
TC0900003299.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900027806.mm.1 |
NM_026917.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027806.mm.1 |
NM_026917.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027806.mm.1 |
TC0900003299.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900027807.mm.1 |
NM_026917.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027807.mm.1 |
NM_026917.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027807.mm.1 |
TC0900003299.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900027811.mm.1 |
NM_026917.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027811.mm.1 |
NM_026917.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027811.mm.1 |
TC0900003299.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900027812.mm.1 |
NM_026917.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027812.mm.1 |
NM_026917.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027812.mm.1 |
TC0900003299.mm.1 |
MTA TC |