Assay Details
| Gene Symbol: Rbfox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110827.2 | NP_001104297.1 | 4 - 5 | 1116 | 116 | |
| NM_001110828.2 | NP_001104298.1 | 3 - 4 | 658 | 116 | ||
| NM_001110829.2 | NP_001104299.1 | 3 - 4 | 658 | 116 | ||
| NM_001110830.2 | NP_001104300.1 | 3 - 4 | 658 | 116 | ||
| NM_001286417.1 | NP_001273346.1 | 3 - 4 | 658 | 116 | ||
| NM_001286419.1 | NP_001273348.1 | 3 - 4 | 655 | 116 | ||
| NM_053104.6 | NP_444334.3 | 4 - 5 | 1116 | 116 | ||
| NM_175387.3 | NP_780596.1 | 4 - 5 | 1116 | 116 | ||
| GenBank mRNA | AF229055.1 | - | 3 - 4 | 601 | 116 | |
| AF387322.1 | - | 3 - 4 | 402 | 116 | ||
| AK044929.1 | - | 3 - 4 | 633 | 116 | ||
| AK077594.1 | - | 4 - 5 | 992 | 116 | ||
| AK139555.1 | - | 5 - 6 | 1086 | 116 | ||
| AY659951.1 | - | 3 - 4 | 402 | 116 | ||
| AY659952.1 | - | 3 - 4 | 402 | 116 | ||
| AY904025.1 | - | 5 - 6 | 552 | 116 | ||
| BC002124.1 | - | 3 - 4 | 496 | 3494630 | 116 | |
| BC027263.1 | - | 4 - 5 | 668 | 3661748 | 116 | |
| DQ017388.1 | - | 3 - 4 | 402 | 116 | ||
| DQ017389.1 | - | 3 - 4 | 399 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbfox2
Genomic Map