Assay Details
| Gene Symbol: Cd209d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130904.2 | NP_570974.1 | 3 - 4 | 239 | 79 | |
| GenBank mRNA | AF373411.1 | - | 3 - 4 | 239 | 79 | |
| AK136586.1 | - | 3 - 4 | 295 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055985 | NM_130904.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055985 | AF373411.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055985 | AK136586.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 15189 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921602 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 192828 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157020 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055985 | NM_130904.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055985 | AF373411.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055985 | AK136586.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615682 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06408348 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157020 | - | - |
| Whole Mouse Genome G4122A | A_52_P401535 | NM_130904.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P401535 | AF373411.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P401535 | AF440280.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P401535 | AK136586.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123517 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157020 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07254816 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07254820 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07254812 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06761025 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07254819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07365303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06743514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07248917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06684580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06684577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06743525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615668 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06797756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06761025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06797755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06797754 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10576812 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576808 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576814 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576815 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576813 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576810 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576809 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576811 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426183_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10576807 | - | - |
| Mouse Genome 430 2.0 Array | 1426183_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1426183_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 170779_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 170779_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800013347.mm.1 | NM_130904.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800013347.mm.1 | NM_130904.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800013347.mm.1 | TC0800001623.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426183_PM_a_at | - | - |
| Mouse Expression Set 430 | 1426183_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426183_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426183_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426183_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426183_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 812277 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2470593 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002470593 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2696291 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2696291 | - | - |
| mouse-6 v1.1 (Illumina) | scl35149.7.1_82-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2470593 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl35149.7.1_82-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2470593 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2696291 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2696291 | - | - |

Cd209d
Genomic Map