Assay Details
| Gene Symbol: Adgre4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139138.3 | NP_631877.2 | 15 - 16 | 1938 | 91 | |
| GenBank mRNA | AF396935.1 | - | 15 - 16 | 1896 | 91 | |
| AK036459.1 | - | 15 - 16 | 1941 | 91 | ||
| AK037483.1 | - | 15 - 16 | 1883 | 91 | ||
| AK155935.1 | - | 15 - 16 | 1886 | 91 | ||
| AY032690.1 | - | 15 - 16 | 1857 | 91 | ||
| BC120538.1 | - | 15 - 16 | 1895 | 40129461 | 91 | |
| BC120540.1 | - | 15 - 16 | 1895 | 40129463 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adgre4
Genomic Map