Assay Details
| Gene Symbol: Acsf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144932.3 | NP_659181.2 | 5 - 6 | 1231 | 78 | |
| GenBank mRNA | AK144352.1 | - | 6 - 7 | 1363 | 78 | |
| BC021611.1 | - | 1 - 2 | 104 | 78 | ||
| BC022709.1 | - | 4 - 5 | 571 | 78 | ||
More Information
Set Membership: |
|

Acsf3
Genomic Map