Assay Details
| Gene Symbol: Cerk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145475.4 | NP_663450.3 | 4 - 5 | 1000 | 75 | |
| GenBank mRNA | AB079067.1 | - | 4 - 5 | 507 | 75 | |
| AK042077.1 | - | 4 - 5 | 591 | 75 | ||
| AK052269.1 | - | 4 - 5 | 766 | 75 | ||
| AK129416.1 | - | 1 - 2 | 142 | 75 | ||
| AK169528.1 | - | 4 - 5 | 585 | 75 | ||
| AK188338.1 | - | 2 - 3 | 230 | 75 | ||
| BC094253.1 | - | 4 - 5 | 999 | 6400872 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cerk
Genomic Map