Assay Details
| Gene Symbol: Rfc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145480.1 | NP_663455.1 | 1 - 2 | 47 | 73 | |
| GenBank mRNA | BC003335.1 | - | 1 - 2 | 47 | 3257208 | 73 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC003335.1,NM_145480.1 transcripts.
Set Membership: |
|

Rfc4
Genomic Map