Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400008805.mm.1 |
NM_145960.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008805.mm.1 |
NM_145960.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008805.mm.1 |
TC1400001102.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400008802.mm.1 |
NM_145960.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008802.mm.1 |
NM_145960.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008802.mm.1 |
TC1400001102.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400008803.mm.1 |
NM_145960.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008803.mm.1 |
NM_145960.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008803.mm.1 |
TC1400001102.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400008804.mm.1 |
NM_145960.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008804.mm.1 |
NM_145960.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008804.mm.1 |
TC1400001102.mm.1 |
MTA TC |