Assay Details
| Gene Symbol: Glis1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147221.2 | NP_671754.2 | 6 - 7 | 1803 | 85 | |
| GenBank mRNA | AF220434.1 | - | 4 - 5 | 1657 | 85 | |
| AF486579.1 | - | 6 - 7 | 1802 | 85 | ||
| AK145342.1 | - | 3 - 4 | 327 | 85 | ||
| BC066157.1 | - | 4 - 5 | 1238 | 30468539 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Glis1
Genomic Map