Assay Details
| Gene Symbol: Slu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148673.3 | NP_683514.2 | 3 - 4 | 350 | 89 | |
| NM_198936.1 | NP_945174.1 | 2 - 3 | 271 | 89 | ||
| GenBank mRNA | AK029117.1 | - | 2 - 3 | 241 | 89 | |
| AK046262.1 | - | 2 - 3 | 242 | 89 | ||
| AK047597.1 | - | 3 - 4 | 338 | 89 | ||
| AK049178.1 | - | 2 - 3 | 240 | 89 | ||
| BC025870.1 | - | 3 - 4 | 329 | 89 | ||
| BC060954.1 | - | 2 - 3 | 239 | 89 | ||
| BC082780.1 | - | 3 - 4 | 330 | 89 | ||
| BC106099.1 | - | 2 - 3 | 252 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slu7
Genomic Map