Assay Details
| Gene Symbol: Padi6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153106.2 | NP_694746.2 | 8 - 9 | 992 | 67 | |
| GenBank mRNA | AF529423.1 | - | 8 - 9 | 968 | 67 | |
| AK139616.1 | - | 8 - 9 | 993 | 67 | ||
| BC053724.1 | - | 8 - 9 | 990 | 30051727 | 67 | |
More Information
Set Membership: |
|

Padi6
Genomic Map