| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100031375.mm.1 |
NM_153137.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031375.mm.1 |
NM_153137.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031375.mm.1 |
TC0100003835.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100031383.mm.1 |
NM_153137.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031383.mm.1 |
NM_153137.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031383.mm.1 |
TC0100003835.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100031381.mm.1 |
NM_153137.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031381.mm.1 |
NM_153137.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031381.mm.1 |
TC0100003835.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100031379.mm.1 |
NM_153137.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031379.mm.1 |
NM_153137.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031379.mm.1 |
TC0100003835.mm.1 |
MTA TC |