Assay Details
Gene Symbol: Slc36a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153139.4 | NP_694779.3 | 9 - 10 | 1142 | 67 | |
GenBank mRNA | AF453743.1 | - | 8 - 9 | 1128 | 67 | |
AY211262.1 | - | 9 - 10 | 1112 | 67 | ||
BC138556.1 | - | 9 - 10 | 1106 | 8861576 | 67 | |
BC138558.1 | - | 9 - 10 | 1106 | 8861578 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
