Assay Details
Gene Symbol: Abca8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153145.4 | NP_694785.3 | 11 - 12 | 1578 | 71 | |
GenBank mRNA | AB255881.1 | - | 10 - 11 | 1497 | 71 | |
AB255882.1 | - | 11 - 12 | 1585 | 71 | ||
AF498360.1 | - | 10 - 11 | 1506 | 71 | ||
AY732492.1 | - | 10 - 11 | 1506 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
