Assay Details
| Gene Symbol: Npb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153288.3 | NP_695020.1 | 1 - 2 | 263 | 90 | |
| GenBank mRNA | AB085945.1 | - | 1 - 2 | 231 | 90 | |
| AK141698.1 | - | 1 - 2 | 264 | 90 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185590 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185588 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185589 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185592 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185593 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185591 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1961089 | NM_153288.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1961089 | AK141698.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952076 | NM_153288.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952076 | AK141698.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952071 | NM_153288.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952071 | AK141698.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 720725 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111021 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1961089 | NM_153288.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1961089 | AK141698.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952071 | NM_153288.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952071 | AK141698.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952076 | NM_153288.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952076 | AK141698.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P300666 | BC134361.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P300666 | NM_153288.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P300666 | AB085945.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P300666 | AK141698.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185589 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185597 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185592 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111021 | - | - |
| Whole Mouse Genome G4122A | A_51_P300666 | BC134361.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P300666 | NM_153288.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P300666 | AB085945.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P300666 | AK141698.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126112 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111021 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09284310 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09284322 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185589 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185593 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185591 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185588 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185590 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09284324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09284322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185594 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10383433 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383431 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383432 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383430 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440240_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10383429 | - | - |
| Mouse Genome 430 2.0 Array | 1440240_at | - | - |
| Mouse Genome 430 2.0 Array | 1440240_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 208990_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 208990_PM_at | - | - |
| Mouse Expression Set 430 | 1440240_at | - | - |
| Mouse Expression Set 430 | 1440240_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440240_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440240_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 691888 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6940632 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006940632 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2635572 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2635572 | - | - |
| mouse-6 v1.1 (Illumina) | scl40627.2.1_25-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6940632 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40627.2.1_25-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6940632 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2635572 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2635572 | - | - |

Npb
Genomic Map