Assay Details
| Gene Symbol: Rtn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007596.2 | NP_001007597.1 | 4 - 5 | 618 | 79 | |
| NM_001286448.1 | NP_001273377.1 | 4 - 5 | 1163 | 79 | ||
| NM_153457.7 | NP_703187.2 | 6 - 7 | 2699 | 79 | ||
| NR_104446.1 | 4 - 5 | 505 | 79 | |||
| GenBank mRNA | AB074899.1 | - | 6 - 7 | 2196 | 79 | |
| AK002997.1 | - | 4 - 5 | 616 | 79 | ||
| AK013731.1 | - | 4 - 5 | 553 | 79 | ||
| AK014358.1 | - | 4 - 5 | 571 | 79 | ||
| AK132364.1 | - | 6 - 7 | 2699 | 79 | ||
| BC027091.1 | - | 4 - 5 | 498 | 79 | ||
| BC030455.1 | - | 6 - 7 | 2251 | 5401661 | 79 | |
| BC053926.1 | - | 6 - 7 | 2327 | 6312477 | 79 | |
| BC058579.1 | - | 6 - 7 | 2361 | 6490092 | 79 | |
| CT010392.1 | - | 6 - 7 | 2196 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rtn1
Genomic Map