Assay Details
| Gene Symbol: Rtn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007596.2 | NP_001007597.1 | 5 - 6 | 663 | 82 | |
| NM_001286448.1 | NP_001273377.1 | 5 - 6 | 1208 | 82 | ||
| NM_153457.7 | NP_703187.2 | 7 - 8 | 2744 | 82 | ||
| NR_104446.1 | 5 - 6 | 550 | 82 | |||
| GenBank mRNA | AB074899.1 | - | 7 - 8 | 2241 | 82 | |
| AK002997.1 | - | 5 - 6 | 661 | 82 | ||
| AK013731.1 | - | 5 - 6 | 598 | 82 | ||
| AK014358.1 | - | 5 - 6 | 616 | 82 | ||
| AK132364.1 | - | 7 - 8 | 2744 | 82 | ||
| BC027091.1 | - | 5 - 6 | 543 | 82 | ||
| BC030455.1 | - | 7 - 8 | 2296 | 5401661 | 82 | |
| BC053926.1 | - | 7 - 8 | 2372 | 6312477 | 82 | |
| BC058579.1 | - | 7 - 8 | 2406 | 6490092 | 82 | |
| CT010392.1 | - | 7 - 8 | 2241 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rtn1
Genomic Map