Assay Details
| Gene Symbol: Fam149a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153535.2 | NP_705763.1 | 9 - 10 | 2279 | 66 | |
| GenBank mRNA | AK034229.1 | - | 9 - 10 | 1506 | 66 | |
| AK038552.1 | - | 9 - 10 | 1494 | 66 | ||
| AK140440.1 | - | 9 - 10 | 2279 | 66 | ||
| AK141607.1 | - | 9 - 10 | 2222 | 66 | ||
| AK220518.1 | - | 9 - 10 | 1515 | 66 | ||
| BC035537.1 | - | 9 - 10 | 2170 | 5369152 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam149a
Genomic Map