Assay Details
| Gene Symbol: E430025E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153548.2 | NP_705776.2 | 19 - 20 | 2618 | 118 | |
| GenBank mRNA | AK088754.1 | - | 4 - 5 | 491 | 118 | |
| AK128970.1 | - | 19 - 20 | 2633 | 118 | ||
| AK129086.1 | - | 347 | 118 | |||
| AK163968.1 | - | 2 - 3 | 242 | 118 | ||
| BC031364.1 | - | 5 - 6 | 632 | 118 | ||
| BC034070.1 | - | 9 - 10 | 1218 | 4217923 | 118 | |
| BC040815.1 | - | 9 - 10 | 1218 | 4217923 | 118 | |
| BC067035.1 | - | 19 - 20 | 2618 | 30532658 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

E430025E21Rik
Genomic Map