Assay Details
| Gene Symbol: E430025E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153548.2 | NP_705776.2 | 27 - 28 | 3571 | 68 | |
| GenBank mRNA | AK088754.1 | - | 12 - 13 | 1444 | 68 | |
| AK128970.1 | - | 27 - 28 | 3586 | 68 | ||
| AK129086.1 | - | 3223 | 68 | |||
| AK163968.1 | - | 10 - 11 | 1195 | 68 | ||
| BC031364.1 | - | 13 - 14 | 1585 | 68 | ||
| BC034070.1 | - | 17 - 18 | 2171 | 4217923 | 68 | |
| BC040815.1 | - | 17 - 18 | 2171 | 4217923 | 68 | |
| BC067035.1 | - | 27 - 28 | 3571 | 30532658 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

E430025E21Rik
Genomic Map