Assay Details
| Gene Symbol: Rnf26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC004739.1 | - | 1 | 951 | 3501511 | 68 |
| BC111036.1 | - | 2 | 889 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1962641 | BC063251.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1962641 | NM_153762.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043330 | BC086757.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043330 | BC111036.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043330 | BC004739.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043330 | BC063251.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043330 | AK085078.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043322 | BC086757.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043322 | BC063251.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043322 | AK208658.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043322 | AK217611.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043322 | AK085078.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043322 | BC086757.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043322 | BC063251.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043322 | AK208658.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043322 | AK217611.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043322 | AK085078.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043330 | BC086757.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043330 | BC111036.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043330 | BC004739.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043330 | BC063251.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043330 | AK085078.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1962641 | BC063251.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1962641 | NM_153762.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26465804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26465816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26465808 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26465815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26465819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26465798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26465806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26465802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26465810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26465822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26465817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26465813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26465801 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26465820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26465805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26465799 | - | - |
| Whole Mouse Genome G4122A | A_51_P201609 | BC086757.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P201609 | BC063251.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P201609 | AK157821.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P201609 | AK033164.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P201609 | NM_153762.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109007 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104159 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07907847 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07907846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26465702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07498475 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07551574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07476747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07732758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07677257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26465771 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10545535 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545536 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545537 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423650_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436824_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10545534 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10592727 | - | - |
| Mouse Genome 430 2.0 Array | 1436824_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1436824_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1423650_at | - | - |
| Mouse Genome 430 2.0 Array | 1423650_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600019541.mm.1 | TC0600002518.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600019543.mm.1 | TC0600002518.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600019542.mm.1 | TC0600002518.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423650_at | - | - |
| Mouse Expression Set 430 | 1423650_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423650_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423650_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423650_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436824_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436824_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423650_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 105340279 | - | - |
| MouseWG-6 v2 Expression BeadChip | 3710292 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105340279 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3710292 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003710292 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2730966 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2730966 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2433047 | - | - |
| mouse-6 v1.1 (Illumina) | scl00213211.1_330-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0213211.1_202-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3710292 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105340279 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00213211.1_330-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3710292 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105340279 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2780420 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2780420 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2433047 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2730966 | - | - |

Rnf26
Genomic Map