Assay Details
| Gene Symbol: Fam20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153782.1 | NP_722477.1 | 3 - 4 | 1186 | 123 | |
| GenBank mRNA | AK142932.1 | - | 3 - 4 | 1210 | 123 | |
| BC011274.1 | - | 3 - 4 | 1092 | 123 | ||
| BC029169.1 | - | 3 - 4 | 1186 | 4947019 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam20a
Genomic Map