Assay Details
| Gene Symbol: Nupl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170591.1 | NP_733479.1 | 2 - 3 | 360 | 67 | |
| GenBank mRNA | AK006062.1 | - | 2 - 3 | 462 | 67 | |
| AK078279.1 | - | 2 - 3 | 408 | 67 | ||
| AK086412.1 | - | 2 - 3 | 428 | 67 | ||
| AK129134.1 | - | 2 - 3 | 397 | 67 | ||
| AK133721.1 | - | 2 - 3 | 371 | 67 | ||
| AK139895.1 | - | 2 - 3 | 490 | 67 | ||
| AK146477.1 | - | 2 - 3 | 360 | 67 | ||
| AK163792.1 | - | 2 - 3 | 389 | 67 | ||
| AK194884.1 | - | 2 - 3 | 174 | 67 | ||
| BC026743.1 | - | 2 - 3 | 391 | 3964978 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nupl1
Genomic Map