Assay Details
| Gene Symbol: Nupl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170591.1 | NP_733479.1 | 3 - 4 | 508 | 73 | |
| GenBank mRNA | AK006062.1 | - | 3 - 4 | 610 | 73 | |
| AK029688.1 | - | 562 | 73 | |||
| AK086412.1 | - | 3 - 4 | 576 | 73 | ||
| AK133721.1 | - | 3 - 4 | 519 | 73 | ||
| AK139895.1 | - | 3 - 4 | 639 | 73 | ||
| AK146477.1 | - | 3 - 4 | 508 | 73 | ||
| AK163792.1 | - | 3 - 4 | 537 | 73 | ||
| BC026743.1 | - | 3 - 4 | 539 | 3964978 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nupl1
Genomic Map