Assay Details
| Gene Symbol: Nupl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170591.1 | NP_733479.1 | 6 - 7 | 780 | 101 | |
| GenBank mRNA | AK029688.1 | - | 834 | 101 | ||
| AK078279.1 | - | 6 - 7 | 828 | 101 | ||
| AK086412.1 | - | 6 - 7 | 848 | 101 | ||
| AK129134.1 | - | 6 - 7 | 778 | 101 | ||
| AK133721.1 | - | 6 - 7 | 791 | 101 | ||
| AK139895.1 | - | 6 - 7 | 911 | 101 | ||
| AK146477.1 | - | 6 - 7 | 780 | 101 | ||
| BC026743.1 | - | 6 - 7 | 811 | 3964978 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nupl1
Genomic Map