Assay Details
| Gene Symbol: Nupl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170591.1 | NP_733479.1 | 10 - 11 | 1216 | 106 | |
| GenBank mRNA | AK006062.1 | - | 10 - 11 | 1319 | 106 | |
| AK029688.1 | - | 1270 | 106 | |||
| AK078279.1 | - | 10 - 11 | 1264 | 106 | ||
| AK086412.1 | - | 10 - 11 | 1284 | 106 | ||
| AK129134.1 | - | 10 - 11 | 1214 | 106 | ||
| AK133721.1 | - | 10 - 11 | 1227 | 106 | ||
| AK139895.1 | - | 10 - 11 | 1347 | 106 | ||
| AK146477.1 | - | 10 - 11 | 1216 | 106 | ||
| BC026743.1 | - | 10 - 11 | 1247 | 3964978 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nupl1
Genomic Map