Assay Details
| Gene Symbol: Nupl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170591.1 | NP_733479.1 | 13 - 14 | 1607 | 65 | |
| GenBank mRNA | AK006062.1 | - | 13 - 14 | 1710 | 65 | |
| AK029688.1 | - | 1661 | 65 | |||
| AK078279.1 | - | 13 - 14 | 1655 | 65 | ||
| AK086412.1 | - | 13 - 14 | 1675 | 65 | ||
| AK129134.1 | - | 13 - 14 | 1605 | 65 | ||
| AK133721.1 | - | 13 - 14 | 1618 | 65 | ||
| AK139895.1 | - | 13 - 14 | 1738 | 65 | ||
| AK146477.1 | - | 13 - 14 | 1607 | 65 | ||
| AK200201.1 | - | 3 - 4 | 335 | 65 | ||
| BC026743.1 | - | 13 - 14 | 1638 | 3964978 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nupl1
Genomic Map