Assay Details
| Gene Symbol: Etv5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023794.2 | NP_076283.2 | 1 - 2 | 57 | 93 | |
| GenBank mRNA | AK018368.1 | - | 1 - 2 | 109 | 93 | |
| AK031452.1 | - | 1 - 2 | 104 | 93 | ||
| AK049234.1 | - | 1 - 2 | 106 | 93 | ||
| AK164091.1 | - | 1 - 2 | 106 | 93 | ||
| AK168881.1 | - | 1 - 2 | 105 | 93 | ||
| BC034680.1 | - | 1 - 2 | 57 | 4036564 | 93 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK018368.1,AK031452.1,AK168881.1,BC034680.1,NM_023794.2 transcripts.
Set Membership: |
|

Etv5
Genomic Map