Assay Details
| Gene Symbol: Etv5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023794.2 | NP_076283.2 | 2 - 3 | 181 | 61 | |
| GenBank mRNA | AK018368.1 | - | 2 - 3 | 233 | 61 | |
| AK031452.1 | - | 2 - 3 | 228 | 61 | ||
| AK049234.1 | - | 2 - 3 | 230 | 61 | ||
| AK080674.1 | - | 1 - 2 | 104 | 61 | ||
| AK164091.1 | - | 2 - 3 | 230 | 61 | ||
| AK168881.1 | - | 2 - 3 | 229 | 61 | ||
| AY004174.1 | - | 1 - 2 | 58 | 61 | ||
| BC034680.1 | - | 2 - 3 | 181 | 4036564 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Etv5
Genomic Map