| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1453449_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10372528 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10366514 |
- |
- |
| Mouse Genome 430 2.0 Array |
1453449_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1453449_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1000010024.mm.1 |
NR_028107.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000010024.mm.1 |
NR_028107.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000010024.mm.1 |
TC1000001338.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000010022.mm.1 |
NR_028107.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000010022.mm.1 |
NR_028107.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000010022.mm.1 |
TC1000001338.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000010023.mm.1 |
NR_028107.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000010023.mm.1 |
NR_028107.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000010023.mm.1 |
TC1000001338.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000010026.mm.1 |
NR_028107.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000010026.mm.1 |
NR_028107.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000010026.mm.1 |
TC1000001338.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1453449_PM_at |
- |
- |
| Mouse Expression Set 430 |
1453449_at |
- |
- |
| Murine Genome U74v2 Set |
169332_at |
- |
- |
| Murine Genome U74v2 Set |
169332_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1453449_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1453449_PM_at |
- |
- |