Assay Details
| Gene Symbol: Yif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026553.4 | NP_080829.1 | 1 - 2 | 209 | 62 | |
| GenBank mRNA | AK011071.1 | - | 1 - 2 | 203 | 62 | |
| AK077329.1 | - | 1 - 2 | 195 | 62 | ||
| AK160409.1 | - | 1 - 2 | 162 | 62 | ||
| BC011117.1 | - | 1 - 2 | 151 | 4190535 | 62 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK077329.1,AK160409.1,BC011117.1,NM_026553.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31926823 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31926822 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P32466 | AK077329.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P32466 | AK160409.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P32466 | BC011117.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P32466 | NM_026553.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P32466 | AK002259.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P32466 | AK077329.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P32466 | AK160409.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P32466 | BC011117.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P32466 | NM_026553.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P32466 | AK002259.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 561738 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588061 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 157933 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100494 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149406 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P32466 | AK077329.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P32466 | AK160409.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P32466 | BC011117.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P32466 | NM_026553.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P32466 | AK002259.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P32466 | AK077329.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P32466 | AK160409.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P32466 | BC011117.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P32466 | NM_026553.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P32466 | AK002259.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926830 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926826 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31926839 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03542375 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149406 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100494 | - | - |
| Whole Mouse Genome G4122A | A_52_P32466 | AK077329.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P32466 | AK160409.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P32466 | BC011117.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P32466 | NM_026553.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P32466 | AK002259.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P32466 | AK077329.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P32466 | AK160409.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P32466 | BC011117.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P32466 | NM_026553.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P32466 | AK002259.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175736 | AK077329.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175736 | AK160409.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175736 | NM_026553.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175736 | AK002259.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101264 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100494 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149406 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14143621 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14143617 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31926822 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31926823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31926842 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14143623 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926840 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926842 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926817 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926811 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14305730 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926831 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14143617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926833 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926826 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926851 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14143639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926816 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926857 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14246474 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926837 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31926813 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10460551 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460549 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460552 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460547 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460545 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460546 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460548 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460550 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460553 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423940_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456134_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423939_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10460544 | - | - |
| Mouse Genome 430 2.0 Array | 1423939_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1456134_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1423940_at | - | - |
| Mouse Genome 430 2.0 Array | 1423939_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1423940_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456134_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68090_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68090_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900000731.mm.1 | NM_026553.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000731.mm.1 | NM_026553.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000731.mm.1 | TC1900000056.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900000729.mm.1 | NM_026553.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000729.mm.1 | NM_026553.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000729.mm.1 | TC1900000056.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900000730.mm.1 | NM_026553.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000730.mm.1 | NM_026553.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000730.mm.1 | TC1900000056.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423940_at | - | - |
| Mouse Expression Set 430 | 1456134_PM_x_at | - | - |
| Mouse Expression Set 430 | 1423939_PM_a_at | - | - |
| Mouse Expression Set 430 | 1423939_a_at | - | - |
| Mouse Expression Set 430 | 1423940_PM_at | - | - |
| Mouse Expression Set 430 | 1456134_x_at | - | - |
| Murine Genome U74v2 Set | 99675_at | - | - |
| Murine Genome U74v2 Set | 99675_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456134_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423940_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423939_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423939_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423940_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456134_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423939_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423939_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423940_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456134_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456134_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423940_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 916529 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5130300 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5130300 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4780079 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005130300 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004780079 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223547 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1235000 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223547 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1235000 | - | - |
| mouse-6 v1.1 (Illumina) | scl000533.1_30-S | - | - |
| mouse-6 v1.1 (Illumina) | scl000532.1_17-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4780079 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5130300 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000532.1_17-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000533.1_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4780079 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5130300 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1235000 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1223547 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2959787 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223547 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1235000 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2959787 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl000533.1_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl000532.1_17-S | - | - |

Yif1a
Genomic Map