| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700020968.mm.1 |
NM_001110824.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020968.mm.1 |
NM_001110824.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020968.mm.1 |
NM_001110825.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020968.mm.1 |
NM_001110825.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020968.mm.1 |
NM_028767.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020968.mm.1 |
NM_028767.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020968.mm.1 |
TC1700002216.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700020973.mm.1 |
NM_001110824.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020973.mm.1 |
NM_001110824.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020973.mm.1 |
NM_001110825.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020973.mm.1 |
NM_001110825.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020973.mm.1 |
NM_028767.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020973.mm.1 |
NM_028767.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020973.mm.1 |
TC1700002216.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700020969.mm.1 |
NM_001110824.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020969.mm.1 |
NM_001110824.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020969.mm.1 |
NM_001110825.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020969.mm.1 |
NM_001110825.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020969.mm.1 |
NM_028767.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020969.mm.1 |
NM_028767.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020969.mm.1 |
TC1700002216.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700020966.mm.1 |
NM_001110824.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020966.mm.1 |
NM_001110824.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020966.mm.1 |
NM_001110825.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020966.mm.1 |
NM_001110825.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020966.mm.1 |
NM_028767.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020966.mm.1 |
NM_028767.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020966.mm.1 |
TC1700002216.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700020974.mm.1 |
NM_001110824.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020974.mm.1 |
NM_001110824.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020974.mm.1 |
NM_001110825.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020974.mm.1 |
NM_001110825.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020974.mm.1 |
NM_028767.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020974.mm.1 |
NM_028767.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020974.mm.1 |
TC1700002216.mm.1 |
MTA TC |