Assay Details
| Gene Symbol: Foxp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110824.1 | NP_001104294.1 | 13 - 14 | 1997 | 66 | |
| NM_001110825.1 | NP_001104295.1 | 13 - 14 | 1961 | 66 | ||
| NM_028767.2 | NP_083043.2 | 13 - 14 | 1958 | 66 | ||
| GenBank mRNA | AB052766.1 | - | 13 - 14 | 1792 | 66 | |
| AK004693.1 | - | 13 - 14 | 1769 | 66 | ||
| AK143194.1 | - | 13 - 14 | 1808 | 66 | ||
| AY135029.1 | - | 13 - 14 | 1785 | 66 | ||
| BC043702.1 | - | 13 - 14 | 1735 | 5351757 | 66 | |
| BC052407.1 | - | 13 - 14 | 1800 | 5716170 | 66 | |
| BC057110.1 | - | 13 - 14 | 1958 | 6417088 | 66 | |
More Information
Set Membership: |
|

Foxp4
Genomic Map