Assay Details
Gene Symbol: Slx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029420.4 | NP_083696.2 | 2 - 3 | 407 | 81 | |
GenBank mRNA | AK010748.1 | - | 2 - 3 | 408 | 81 | |
AK010828.1 | - | 2 - 3 | 371 | 81 | ||
AK014753.1 | - | 2 - 3 | 264 | 81 | ||
AK049139.1 | - | 2 - 3 | 408 | 81 | ||
BC055909.1 | - | 2 - 3 | 359 | 81 | ||
BC145688.1 | - | 2 - 3 | 272 | 40130991 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
