Assay Details
| Gene Symbol: Asl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133768.4 | NP_598529.1 | 5 - 6 | 429 | 73 | |
| GenBank mRNA | AK076453.1 | - | 5 - 6 | 444 | 73 | |
| AK138423.1 | - | 5 - 6 | 426 | 73 | ||
| AK140160.1 | - | 5 - 6 | 444 | 73 | ||
| AK148764.1 | - | 5 - 6 | 413 | 73 | ||
| AK168739.1 | - | 5 - 6 | 424 | 73 | ||
| BC016670.1 | - | 5 - 6 | 429 | 2648753 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asl
Genomic Map