Assay Details
| Gene Symbol: Asl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133768.4 | NP_598529.1 | 13 - 14 | 1055 | 89 | |
| GenBank mRNA | AK010922.1 | - | 13 - 14 | 1067 | 89 | |
| AK138423.1 | - | 13 - 14 | 1052 | 89 | ||
| AK140160.1 | - | 13 - 14 | 1070 | 89 | ||
| AK148764.1 | - | 13 - 14 | 1039 | 89 | ||
| AK168739.1 | - | 13 - 14 | 1050 | 89 | ||
| AK185794.1 | - | 1 - 2 | 52 | 89 | ||
| AK215777.1 | - | 1 - 2 | 51 | 89 | ||
| BC016670.1 | - | 13 - 14 | 1055 | 2648753 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asl
Genomic Map