Assay Details
| Gene Symbol: Zfp410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144833.3 | NP_659082.1 | 10 - 11 | 1732 | 83 | |
| NR_045546.1 | 10 - 11 | 1700 | 83 | |||
| NR_045615.1 | 8 - 9 | 1416 | 83 | |||
| GenBank mRNA | AF295806.1 | - | 10 - 11 | 1494 | 83 | |
| AK051939.1 | - | 8 - 9 | 1132 | 83 | ||
| AK132345.1 | - | 10 - 11 | 1471 | 83 | ||
| AK181763.1 | - | 1 - 2 | 142 | 83 | ||
| BC021528.1 | - | 10 - 11 | 1487 | 5353673 | 83 | |
| BC056957.1 | - | 10 - 11 | 1429 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp410
Genomic Map