Assay Details
| Gene Symbol: Emilin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145158.3 | NP_660140.1 | 5 - 6 | 2836 | 142 | |
| GenBank mRNA | AF468645.1 | - | 5 - 6 | 2787 | 142 | |
| AK154230.1 | - | 5 - 6 | 2806 | 142 | ||
| AK155037.1 | - | 5 - 6 | 2816 | 142 | ||
| AK155306.1 | - | 5 - 6 | 2830 | 142 | ||
| AK155366.1 | - | 5 - 6 | 2817 | 142 | ||
| AK155554.1 | - | 5 - 6 | 2832 | 142 | ||
| AK155878.1 | - | 5 - 6 | 2835 | 142 | ||
| AK155879.1 | - | 5 - 6 | 2830 | 143 | ||
| AK155909.1 | - | 5 - 6 | 2836 | 142 | ||
| AK155913.1 | - | 5 - 6 | 2806 | 142 | ||
| AK155918.1 | - | 5 - 6 | 2864 | 142 | ||
| AK170081.1 | - | 5 - 6 | 2830 | 142 | ||
| AK170391.1 | - | 5 - 6 | 2806 | 142 | ||
| AK170801.1 | - | 5 - 6 | 2837 | 142 | ||
| AK170822.1 | - | 5 - 6 | 2858 | 142 | ||
| AK170953.1 | - | 5 - 6 | 2852 | 142 | ||
| AK171015.1 | - | 5 - 6 | 2858 | 142 | ||
| AK171020.1 | - | 5 - 6 | 2824 | 142 | ||
| AK171303.1 | - | 5 - 6 | 2808 | 142 | ||
| AK171309.1 | - | 5 - 6 | 2833 | 142 | ||
| AK171579.1 | - | 5 - 6 | 2806 | 143 | ||
| AK171592.1 | - | 5 - 6 | 2806 | 142 | ||
| AK171616.1 | - | 5 - 6 | 2807 | 142 | ||
| BC053753.1 | - | 5 - 6 | 3120 | 6510220 | 142 | |
| GQ872457.1 | - | 5 - 6 | 2731 | 142 | ||
| GQ872460.1 | - | 5 - 6 | 2731 | 142 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Emilin2
Genomic Map